Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
Chromosome Location
Chr.6: 32290523 - 32371912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006781.4 NP_006772.3 20 - 21 811 103
XM_017010185.1 XP_016865674.1 21 - 22 836 103
GenBank mRNA BC034774.1 - 20 - 21 778 103

View Full Product Details