Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
Chromosome Location
Chr.6: 32290523 - 32371912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286474.1 NP_001273403.1 3 - 4 342 147
NM_001286475.1 NP_001273404.1 3 - 4 342 147
NM_006781.4 NP_006772.3 3 - 4 342 147
GenBank mRNA AK225742.1 - 3 - 4 333 147
AK302176.1 - 3 - 4 302 147
AK310230.1 - 3 - 4 331 147
BC034774.1 - 3 - 4 309 147
DC399589.1 - 3 - 4 343 147

View Full Product Details