Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
Chromosome Location
Chr.6: 32290523 - 32371912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286474.1 NP_001273403.1 4 - 5 373 126
NM_001286475.1 NP_001273404.1 4 - 5 373 105
NM_006781.4 NP_006772.3 4 - 5 373 126
GenBank mRNA AK225742.1 - 4 - 5 364 126
AK302176.1 - 4 - 5 333 105
AK310230.1 - 4 - 5 362 105
BC034774.1 - 4 - 5 340 126
DC399589.1 - 4 - 5 374 126
U60665.1 - 4 - 5 403 126

View Full Product Details