The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042450.2 | NP_001035915.1 | 2 - 3 | 219 | 87 |
NM_001270648.1 | NP_001257577.1 | 2 - 3 | 219 | 87 | |
NM_001270649.1 | NP_001257578.1 | 2 - 3 | 219 | 87 | |
NM_001282417.1 | NP_001269346.1 | 2 - 3 | 250 | 87 | |
NM_152351.4 | NP_689564.3 | 2 - 3 | 219 | 87 | |
XM_011523652.1 | XP_011521954.1 | 2 - 3 | 375 | 87 | |
XM_011523653.2 | XP_011521955.1 | 2 - 3 | 380 | 87 | |
XM_017024191.1 | XP_016879680.1 | 2 - 3 | 376 | 87 | |
GenBank mRNA | AK057946.1 | - | 2 - 3 | 219 | 87 |
AK298345.1 | - | 2 - 3 | 205 | 87 | |
BC034380.1 | - | 2 - 3 | 164 | 87 | |
BC039868.1 | - | 2 - 3 | 267 | 87 | |
BC062617.1 | - | 2 - 3 | 173 | 87 | |
BP332461.1 | - | 2 - 3 | 219 | 87 | |
DR003789.1 | - | 2 - 3 | 219 | 87 |