The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002759.1 | NP_001002759.1 | 3 - 3 | 449 | 103 |
NM_145247.4 | NP_660290.3 | 3 - 3 | 520 | 103 | |
XM_005269521.3 | XP_005269578.1 | 2 - 2 | 1093 | 103 | |
XM_017015672.1 | XP_016871161.1 | 3 - 3 | 487 | 103 | |
GenBank mRNA | AK123954.1 | - | 2 - 2 | 2087 | 103 |
AK291184.1 | - | 2 - 2 | 1143 | 103 | |
BC020892.2 | - | 2 - 2 | 424 | 103 | |
BC047587.2 | - | 1 - 1 | 1051 | 103 | |
BC140835.1 | - | 3 - 3 | 481 | 103 |