Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERF1A
Entrez Gene ID
8293
Gene Name
small EDRK-rich factor 1A
Gene Aliases
4F5, FAM2A, H4F5, SERF1, SMAM1
Chromosome Location
Chr.5: 70900663 - 70918530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022968.1 NP_075257.1 3 - 3 545 153
GenBank mRNA AF073518.1 - 3 - 3 512 153
BC021174.1 - 3 - 3 544 153
BX107837.1 - 3 - 3 470 153

Gene Symbol
SERF1B
Entrez Gene ID
728492
Gene Name
small EDRK-rich factor 1B
Gene Aliases
FAM2B, H4F5C, h4F5
Chromosome Location
Chr.5: 70025251 - 70043108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178087.1 NP_001171558.1 3 - 3 539 153
GenBank mRNA CD679521.1 - 3 - 3 419 153

View Full Product Details