The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002568.3 | NP_002559.2 | 1 - 1 | 47 | 123 |
XM_005250861.2 | XP_005250918.1 | 1 - 1 | 958 | 123 | |
GenBank mRNA | AK130523.1 | - | 1 - 1 | 47 | 123 |
AK303120.1 | - | 1 - 1 | 47 | 123 | |
BC023217.1 | - | 1 - 1 | 26 | 123 | |
BC023520.2 | - | 1 - 1 | 26 | 123 | |
BC041863.1 | - | 1 - 1 | 671 | 123 | |
Y00345.1 | - | 1 - 1 | 46 | 123 |