Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DKKL1
Entrez Gene ID
27120
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
CT34, SGY, SGY-1, SGY1
Chromosome Location
Chr.19: 49360520 - 49375132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197301.1 NP_001184230.1 1 - 1 165 55
NM_014419.3 NP_055234.1 1 - 1 165 55
GenBank mRNA AI879324.1 - 1 - 1 356 55
AY358922.1 - 1 - 1 165 55
BC030581.2 - 1 - 1 45 55

View Full Product Details