The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003577.2 | NP_003568.2 | 1 - 2 | 566 | 177 |
GenBank mRNA | AB011076.2 | - | 1 - 2 | 567 | 177 |
AY606112.1 | - | 1 - 2 | 566 | 177 |