The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003017.4 | NP_003008.1 | 6 - 6 | 1826 | 93 |
NR_036610.1 | 7 - 7 | 2282 | 93 | ||
GenBank mRNA | AF107405.1 | - | 6 - 6 | 1749 | 93 |