The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177400.2 | NP_796374.1 | 3 - 3 | 839 | 89 |
XM_017016789.1 | XP_016872278.1 | 3 - 3 | 833 | 89 | |
GenBank mRNA | BC101635.1 | - | 3 - 3 | 839 | 89 |
BC104823.1 | - | 3 - 3 | 839 | 89 | |
BF509028.1 | - | 3 - 3 | 469 | 89 |