The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000109.3 | NP_000100.2 | 75 - 76 | 11121 | 73 |
NM_004006.2 | NP_003997.1 | 75 - 76 | 11045 | 73 | |
NM_004009.3 | NP_004000.1 | 75 - 76 | 11052 | 73 | |
NM_004010.3 | NP_004001.1 | 75 - 76 | 11135 | 73 | |
NM_004011.3 | NP_004002.2 | 47 - 48 | 6823 | 73 | |
NM_004012.3 | NP_004003.1 | 47 - 48 | 6966 | 73 | |
NM_004013.2 | NP_004004.1 | 32 - 33 | 4462 | 73 | |
NM_004014.2 | NP_004005.1 | 21 - 22 | 2675 | 73 | |
NM_004015.2 | NP_004006.1 | 14 - 15 | 1675 | 73 | |
NM_004016.2 | NP_004007.1 | 14 - 15 | 1675 | 73 | |
NM_004017.2 | NP_004008.1 | 13 - 14 | 1636 | 73 | |
NM_004018.2 | NP_004009.1 | 13 - 14 | 1636 | 73 | |
NM_004020.3 | NP_004011.2 | 28 - 29 | 4132 | 73 | |
NM_004021.2 | NP_004012.1 | 32 - 33 | 4462 | 73 | |
NM_004022.2 | NP_004013.1 | 31 - 32 | 4423 | 73 | |
NM_004023.2 | NP_004014.1 | 28 - 29 | 4132 | 73 | |
XM_006724468.2 | XP_006724531.1 | 75 - 76 | 10998 | 73 | |
XM_006724469.3 | XP_006724532.1 | 75 - 76 | 10972 | 73 | |
XM_006724470.3 | XP_006724533.1 | 74 - 75 | 10959 | 73 | |
XM_006724473.2 | XP_006724536.1 | 74 - 75 | 10860 | 73 | |
XM_006724474.3 | XP_006724537.1 | 71 - 72 | 10668 | 73 | |
XM_006724475.2 | XP_006724538.1 | 71 - 72 | 10668 | 73 | |
XM_011545467.1 | XP_011543769.1 | 74 - 75 | 10875 | 73 | |
XM_017029328.1 | XP_016884817.1 | 74 - 75 | 10959 | 73 | |
XM_017029331.1 | XP_016884820.1 | 35 - 36 | 5574 | 73 | |
GenBank mRNA | AB208836.1 | - | 28 - 29 | 3870 | 73 |
AK299936.1 | - | 10 - 11 | 1387 | 73 | |
BC028720.1 | - | 14 - 15 | 1726 | 73 | |
BC070078.1 | - | 13 - 14 | 1637 | 73 | |
BC094758.1 | - | 14 - 15 | 1689 | 73 | |
BC127103.1 | - | 29 - 30 | 3986 | 73 | |
BC150141.1 | - | 29 - 30 | 3986 | 73 | |
KU178030.1 | - | 14 - 15 | 1597 | 73 | |
KU178031.1 | - | 14 - 15 | 1597 | 73 | |
KU178032.1 | - | 10 - 11 | 1267 | 73 | |
M18533.1 | - | 75 - 76 | 11009 | 73 | |
M92650.1 | - | 13 - 14 | 1610 | 73 | |
X14298.1 | - | 75 - 76 | 10899 | 73 |