Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 16 - 17 1967 112
NM_001144770.1 NP_001138242.1 14 - 15 1638 112
NM_001723.5 NP_001714.1 3 - 4 522 112
NM_015548.4 NP_056363.2 3 - 4 522 112
NM_183380.3 NP_899236.1 13 - 14 1912 112
XM_005249310.3 XP_005249367.1 17 - 18 2061 112
XM_005249315.3 XP_005249372.1 16 - 17 2008 112
XM_005249316.2 XP_005249373.1 9 - 10 2284 112
XM_005249318.2 XP_005249375.1 15 - 16 1722 112
XM_005249319.2 XP_005249376.1 14 - 15 1623 112
XM_005249320.4 XP_005249377.1 14 - 15 1679 112
XM_005249322.4 XP_005249379.1 14 - 15 1574 112
XM_005249323.3 XP_005249380.1 13 - 14 1907 112
XM_005249324.2 XP_005249381.1 3 - 4 519 112
XM_011514824.2 XP_011513126.1 16 - 17 1972 112
XM_011514825.2 XP_011513127.1 16 - 17 1968 112
XM_011514826.2 XP_011513128.1 13 - 14 1907 112
XM_017011205.1 XP_016866694.1 17 - 18 2071 112
XM_017011206.1 XP_016866695.1 17 - 18 2071 112
XM_017011207.1 XP_016866696.1 17 - 18 2008 112
XM_017011208.1 XP_016866697.1 17 - 18 2071 112
XM_017011209.1 XP_016866698.1 17 - 18 2071 112
XM_017011210.1 XP_016866699.1 17 - 18 2071 112
XM_017011211.1 XP_016866700.1 17 - 18 2071 112
XM_017011212.1 XP_016866701.1 17 - 18 2071 112
XM_017011213.1 XP_016866702.1 17 - 18 2071 112
XM_017011214.1 XP_016866703.1 17 - 18 2071 112
XM_017011215.1 XP_016866704.1 17 - 18 2071 112
XM_017011216.1 XP_016866705.1 17 - 18 2071 112
XM_017011217.1 XP_016866706.1 14 - 15 1652 112
XM_017011219.1 XP_016866708.1 17 - 18 2067 112
XM_017011220.1 XP_016866709.1 16 - 17 2010 112
XM_017011221.1 XP_016866710.1 17 - 18 2067 112
XM_017011222.1 XP_016866711.1 9 - 10 2226 112
XM_017011223.1 XP_016866712.1 17 - 18 2067 112
XM_017011224.1 XP_016866713.1 13 - 14 1907 112
XM_017011225.1 XP_016866714.1 17 - 18 2081 112
GenBank mRNA AF400226.1 - 3 - 4 414 112
AF400227.1 - 3 - 4 414 112
AK294830.1 - 13 - 14 1520 112
AK295864.1 - 9 - 10 1894 112
AK299898.1 - 14 - 15 1638 112
BC065536.1 - 3 - 4 486 112
DA786051.1 - 4 - 5 409 112
L11690.1 - 3 - 4 516 112
M69225.1 - 3 - 4 452 112

View Full Product Details