The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002862.3 | NP_002853.2 | 19 - 20 | 2489 | 93 |
GenBank mRNA | AB209080.1 | - | 19 - 20 | 93 | |
AF432222.1 | - | 4 - 5 | 457 | 93 | |
AK299697.1 | - | 19 - 20 | 93 | ||
BC017045.2 | - | 19 - 20 | 2464 | 93 | |
BC030795.2 | - | 19 - 20 | 2465 | 93 | |
J03544.1 | - | 19 - 20 | 2459 | 93 | |
U47025.1 | - | 19 - 20 | 2414 | 93 |