The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165412.1 | NP_001158884.1 | 20 - 21 | 2813 | 117 |
NM_001319226.1 | NP_001306155.1 | 20 - 21 | 2623 | 117 | |
NM_003998.3 | NP_003989.2 | 20 - 21 | 2816 | 117 | |
XM_011532006.2 | XP_011530308.1 | 21 - 22 | 2583 | 117 | |
XM_011532008.2 | XP_011530310.1 | 18 - 19 | 2197 | 117 | |
GenBank mRNA | AK122850.1 | - | 13 - 14 | 2945 | 117 |
AK291450.1 | - | 20 - 21 | 2477 | 117 | |
BC033210.1 | - | 8 - 9 | 1094 | 117 | |
BC051765.1 | - | 20 - 21 | 2816 | 117 | |
CR749522.1 | - | 20 - 21 | 2573 | 117 | |
M55643.1 | - | 20 - 21 | 2530 | 117 | |
M58603.1 | - | 20 - 21 | 2743 | 117 |