Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HNF4A
- Entrez Gene ID
-
3172
- Gene Name
- hepatocyte nuclear factor 4 alpha
- Gene Aliases
-
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
- Chromosome Location
-
Chr.20: 44355801 - 44432845 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK096973.1 |
- |
1 - 1 |
1256 |
118 |
View Full Product Details