Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236409135-236409374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485606 c.76G>A p.E26K Chr.1: 236409230-236409230 on Build GRCh38
6651547 c.79C>T p.P27S Chr.1: 236409233-236409233 on Build GRCh38
6651548 c.49C>T p.P17S Chr.1: 236409233-236409233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758595184 Chr.1: 236409283-236409283 on Build GRCh38
rs773039572 Chr.1: 236409215-236409215 on Build GRCh38
rs561106854 Chr.1: 236409184-236409184 on Build GRCh38
rs374625285 Chr.1: 236409255-236409255 on Build GRCh38
rs369439024 Chr.1: 236409208-236409208 on Build GRCh38
rs765501920 Chr.1: 236409282-236409282 on Build GRCh38
rs149429886 Chr.1: 236409281-236409281 on Build GRCh38
rs767234735 Chr.1: 236409169-236409169 on Build GRCh38
rs377143490 Chr.1: 236409198-236409198 on Build GRCh38
rs756282604 Chr.1: 236409172-236409172 on Build GRCh38
rs749338738 Chr.1: 236409181-236409181 on Build GRCh38
rs568769533 Chr.1: 236409325-236409325 on Build GRCh38
rs767467962 Chr.1: 236409277-236409277 on Build GRCh38
rs1043481004 Chr.1: 236409299-236409299 on Build GRCh38
rs770542352 Chr.1: 236409226-236409226 on Build GRCh38
rs759461234 Chr.1: 236409269-236409269 on Build GRCh38
rs370449930 Chr.1: 236409300-236409300 on Build GRCh38
rs200569815 Chr.1: 236409237-236409237 on Build GRCh38
rs769530943 Chr.1: 236409209-236409209 on Build GRCh38
rs144815146 Chr.1: 236409316-236409316 on Build GRCh38
rs762357435 Chr.1: 236409218-236409218 on Build GRCh38
rs879255553 Chr.1: 236409275-236409275 on Build GRCh38
rs565166177 Chr.1: 236409262-236409262 on Build GRCh38
rs754143479 Chr.1: 236409294-236409294 on Build GRCh38
rs1054925527 Chr.1: 236409342-236409342 on Build GRCh38
rs1009732221 Chr.1: 236409166-236409166 on Build GRCh38
rs757277070 Chr.1: 236409296-236409296 on Build GRCh38
rs746451148 Chr.1: 236409187-236409187 on Build GRCh38
rs778980961 Chr.1: 236409298-236409298 on Build GRCh38
rs772283262 Chr.1: 236409319-236409319 on Build GRCh38
rs760506554 Chr.1: 236409284-236409284 on Build GRCh38
rs757403340 Chr.1: 236409183-236409183 on Build GRCh38
rs752504949 Chr.1: 236409170-236409170 on Build GRCh38
rs669710 Chr.1: 236409175-236409175 on Build GRCh38
rs773740123 Chr.1: 236409250-236409250 on Build GRCh38
rs746034923 Chr.1: 236409212-236409212 on Build GRCh38
rs758889519 Chr.1: 236409314-236409314 on Build GRCh38
rs540466096 Chr.1: 236409259-236409259 on Build GRCh38

View Full Product Details