Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCARF1
Entrez Gene ID
8578
Gene Name
scavenger receptor class F member 1
Gene Aliases
SREC, SREC-I, SREC1
Chromosome Location
Chr.17: 1633858 - 1645789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003693.3 NP_003684.2 2 - 3 259 114
NM_145350.2 NP_663325.1 2 - 3 259 114
NR_028075.2 2 - 3 259 114
NR_102409.1 2 - 3 296 114
GenBank mRNA AB052947.1 - 2 - 3 193 114
AB052948.1 - 2 - 3 226 114
AB052949.1 - 2 - 3 191 114
AB052950.1 - 2 - 3 183 114
AK291810.1 - 2 - 3 202 114
AK310174.1 - 2 - 3 219 114
BC039735.1 - 2 - 3 227 114
D63483.1 - 2 - 3 169 114
D86864.1 - 2 - 3 193 114

View Full Product Details