The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003404.4 | NP_003395.1 | 5 - 6 | 974 | 132 |
NM_139323.3 | NP_647539.1 | 4 - 5 | 879 | 132 | |
XM_017028039.1 | XP_016883528.1 | 5 - 6 | 778 | 132 | |
GenBank mRNA | AK292717.1 | - | 5 - 6 | 867 | 132 |
AL834170.1 | - | 4 - 5 | 758 | 132 | |
BC001359.2 | - | 4 - 5 | 769 | 132 | |
EU794580.1 | - | 5 - 6 | 870 | 132 | |
X57346.1 | - | 5 - 6 | 960 | 132 |