The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290048.1 | NP_001276977.1 | 12 - 13 | 1829 | 109 |
NM_015459.4 | NP_056274.3 | 12 - 13 | 1931 | 109 | |
XM_006718493.1 | XP_006718556.1 | 11 - 12 | 1756 | 109 | |
GenBank mRNA | AK023383.1 | - | 12 - 13 | 1536 | 109 |
AK097588.1 | - | 14 - 15 | 1871 | 109 | |
AK301910.1 | - | 12 - 13 | 1767 | 109 | |
AL117600.1 | - | 12 - 13 | 1578 | 109 | |
BC077727.1 | - | 12 - 13 | 1822 | 109 | |
BX642720.1 | - | 1 - 2 | 171 | 109 | |
CR936784.1 | - | 12 - 13 | 1814 | 109 |