Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 79 - 80 14440 76
NM_001144770.1 NP_001138242.1 77 - 78 14111 76
NM_015548.4 NP_056363.2 66 - 67 12995 76
NM_183380.3 NP_899236.1 76 - 77 14385 76
XM_005249310.3 XP_005249367.1 85 - 86 20792 76
XM_005249315.3 XP_005249372.1 84 - 85 20739 76
XM_005249316.2 XP_005249373.1 77 - 78 21015 76
XM_005249318.2 XP_005249375.1 83 - 84 20453 76
XM_005249319.2 XP_005249376.1 82 - 83 20354 76
XM_005249320.4 XP_005249377.1 82 - 83 20410 76
XM_005249322.4 XP_005249379.1 82 - 83 20305 76
XM_005249323.3 XP_005249380.1 81 - 82 20638 76
XM_005249324.2 XP_005249381.1 71 - 72 19250 76
XM_011514824.2 XP_011513126.1 84 - 85 20703 76
XM_011514825.2 XP_011513127.1 79 - 80 14441 76
XM_011514826.2 XP_011513128.1 80 - 81 20311 76
XM_017011205.1 XP_016866694.1 85 - 86 20802 76
XM_017011206.1 XP_016866695.1 85 - 86 20802 76
XM_017011207.1 XP_016866696.1 85 - 86 20739 76
XM_017011208.1 XP_016866697.1 85 - 86 20802 76
XM_017011209.1 XP_016866698.1 85 - 86 20802 76
XM_017011210.1 XP_016866699.1 85 - 86 20802 76
XM_017011211.1 XP_016866700.1 85 - 86 20802 76
XM_017011212.1 XP_016866701.1 84 - 85 20568 76
XM_017011213.1 XP_016866702.1 84 - 85 20475 76
XM_017011214.1 XP_016866703.1 84 - 85 20475 76
XM_017011215.1 XP_016866704.1 84 - 85 20475 76
XM_017011216.1 XP_016866705.1 84 - 85 20475 76
XM_017011217.1 XP_016866706.1 82 - 83 20383 76
XM_017011218.1 XP_016866707.1 47 - 48 16411 76
XM_017011219.1 XP_016866708.1 80 - 81 14540 76
XM_017011220.1 XP_016866709.1 79 - 80 14483 76
XM_017011221.1 XP_016866710.1 79 - 80 14306 76
XM_017011222.1 XP_016866711.1 72 - 73 14699 76
XM_017011223.1 XP_016866712.1 79 - 80 14213 76
XM_017011224.1 XP_016866713.1 75 - 76 14053 76
GenBank mRNA AB018271.2 - 14 - 15 2285 76
AF400226.1 - 66 - 67 12887 76

View Full Product Details