Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50579056-50579307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6876229 c.1184C>T p.S395F Chr.22: 50579185-50579185 on Build GRCh38
1417175 c.1114-1G>T p.? Chr.22: 50579256-50579256 on Build GRCh38
1682380 c.1121C>T p.A374V Chr.22: 50579248-50579248 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886876527 Chr.22: 50579111-50579111 on Build GRCh38
rs769298325 Chr.22: 50579236-50579236 on Build GRCh38
rs886057638 Chr.22: 50579258-50579258 on Build GRCh38
rs1048502182 Chr.22: 50579096-50579096 on Build GRCh38
rs778061654 Chr.22: 50579275-50579275 on Build GRCh38
rs759847956 Chr.22: 50579256-50579256 on Build GRCh38
rs766848672 Chr.22: 50579240-50579240 on Build GRCh38
rs980683552 Chr.22: 50579196-50579196 on Build GRCh38
rs994057235 Chr.22: 50579080-50579080 on Build GRCh38
rs969288304 Chr.22: 50579195-50579195 on Build GRCh38
rs551576081 Chr.22: 50579175-50579175 on Build GRCh38
rs758563051 Chr.22: 50579248-50579249 on Build GRCh38
rs762677 Chr.22: 50579284-50579284 on Build GRCh38
rs761473672 Chr.22: 50579223-50579223 on Build GRCh38
rs772705206 Chr.22: 50579239-50579239 on Build GRCh38
rs17001634 Chr.22: 50579085-50579085 on Build GRCh38
rs533323083 Chr.22: 50579160-50579160 on Build GRCh38
rs768460701 Chr.22: 50579205-50579205 on Build GRCh38
rs752796169 Chr.22: 50579134-50579134 on Build GRCh38
rs781238750 Chr.22: 50579135-50579135 on Build GRCh38
rs140289037 Chr.22: 50579212-50579212 on Build GRCh38
rs151123092 Chr.22: 50579261-50579261 on Build GRCh38
rs773322949 Chr.22: 50579278-50579278 on Build GRCh38
rs746752350 Chr.22: 50579202-50579202 on Build GRCh38
rs772125270 Chr.22: 50579188-50579188 on Build GRCh38
rs767871830 Chr.22: 50579259-50579259 on Build GRCh38
rs756289571 Chr.22: 50579269-50579269 on Build GRCh38
rs758453999 Chr.22: 50579282-50579282 on Build GRCh38
rs375459778 Chr.22: 50579287-50579287 on Build GRCh38
rs1004350925 Chr.22: 50579114-50579114 on Build GRCh38
rs527814172 Chr.22: 50579279-50579279 on Build GRCh38
rs752921190 Chr.22: 50579148-50579148 on Build GRCh38
rs751958642 Chr.22: 50579248-50579248 on Build GRCh38
rs970008885 Chr.22: 50579251-50579251 on Build GRCh38
rs372529332 Chr.22: 50579235-50579235 on Build GRCh38
rs371344816 Chr.22: 50579154-50579154 on Build GRCh38
rs780047741 Chr.22: 50579193-50579193 on Build GRCh38
rs756167681 Chr.22: 50579159-50579159 on Build GRCh38

View Full Product Details