Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB41
Entrez Gene ID
360023
Gene Name
zinc finger and BTB domain containing 41
Gene Aliases
FRBZ1, ZNF924
Chromosome Location
Chr.1: 197153682 - 197200542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194314.2 NP_919290.2 1 - 2 1192 118
NR_135153.1 1 - 2 1192 118
XM_011509507.2 XP_011507809.1 1 - 2 1171 118
XM_017001205.1 XP_016856694.1 1 - 2 1171 118
GenBank mRNA AK128549.1 - 1 - 2 1171 118
AY163816.2 - 1 - 2 1192 118
BC132818.1 - 1 - 2 1156 118
BC136600.1 - 1 - 2 1156 118

View Full Product Details