The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005039.3 | NP_005030.2 | 3 - 4 | 1061 | 121 |
NM_199353.2 | NP_955385.1 | 4 - 5 | 662 | 121 | |
NM_199354.2 | NP_955386.1 | 4 - 5 | 602 | 121 | |
GenBank mRNA | K03204.1 | - | 3 - 4 | 1061 | 121 |
K03205.1 | - | 4 - 5 | 650 | 121 | |
K03206.1 | - | 4 - 5 | 598 | 121 |