The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032588.3 | NP_115977.2 | 1 - 2 | 299 | 55 |
XM_017002559.1 | XP_016858048.1 | 1 - 2 | 299 | 55 | |
GenBank mRNA | AF353673.1 | - | 1 - 2 | 278 | 55 |
AF361946.1 | - | 1 - 2 | 1103 | 55 | |
AJ276484.1 | - | 1 - 2 | 282 | 55 | |
AK056942.1 | - | 1 - 2 | 295 | 55 | |
AK297820.1 | - | 1 - 2 | 295 | 55 | |
BC080529.1 | - | 1 - 2 | 277 | 55 | |
BT007373.1 | - | 1 - 2 | 159 | 55 | |
DA900348.1 | - | 1 - 2 | 299 | 55 |