The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195685.1 | NP_001182614.1 | 4 - 5 | 633 | 77 |
NM_194317.3 | NP_919298.1 | 4 - 5 | 623 | 77 | |
GenBank mRNA | AK124733.1 | - | 4 - 5 | 633 | 77 |
AK309248.1 | - | 5 - 6 | 787 | 77 | |
AL831852.1 | - | 1 - 2 | 67 | 77 | |
AY358623.1 | - | 4 - 5 | 539 | 77 | |
BC032836.1 | - | 7 - 8 | 1155 | 77 | |
BC047013.1 | - | 5 - 6 | 653 | 77 |