Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HLA-DRB1
- Entrez Gene ID
-
3123
- Gene Name
- major histocompatibility complex, class II, DR beta 1
- Gene Aliases
-
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
- Chromosome Location
-
Chr.6: 32578769 - 32589836 on Build GRCh38
- Gene Symbol
- HLA-DRB4
- Entrez Gene ID
-
3126
- Gene Name
- major histocompatibility complex, class II, DR beta 4
- Gene Aliases
-
DR-4, DR4, DRB4, HLA-DR4B
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK225683.1 |
- |
1 - 1 |
43 |
115 |
AK225690.1 |
- |
1 - 1 |
43 |
115 |
AK291499.1 |
- |
1 - 1 |
43 |
115 |
AK293020.1 |
- |
1 - 1 |
84 |
115 |
View Full Product Details