The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019050.2 | NP_061923.2 | 18 - 18 | 4928 | 131 |
XM_005263073.3 | XP_005263130.1 | 17 - 17 | 4802 | 131 | |
XM_005263075.2 | XP_005263132.1 | 17 - 17 | 4817 | 131 | |
XM_005263076.2 | XP_005263133.1 | 17 - 17 | 4814 | 131 | |
XM_005263077.3 | XP_005263134.1 | 17 - 17 | 4870 | 131 | |
XM_006714240.1 | XP_006714303.1 | 16 - 16 | 4003 | 131 | |
XM_011532037.2 | XP_011530339.1 | 18 - 18 | 4980 | 131 | |
XM_011532038.2 | XP_011530340.1 | 17 - 17 | 4257 | 131 | |
XM_017008312.1 | XP_016863801.1 | 19 - 19 | 5094 | 131 | |
XM_017008313.1 | XP_016863802.1 | 18 - 18 | 4779 | 131 | |
XM_017008314.1 | XP_016863803.1 | 18 - 18 | 4778 | 131 | |
XM_017008315.1 | XP_016863804.1 | 18 - 18 | 4848 | 131 | |
XM_017008316.1 | XP_016863805.1 | 16 - 16 | 4666 | 131 | |
XM_017008317.1 | XP_016863806.1 | 16 - 16 | 4694 | 131 | |
XM_017008318.1 | XP_016863807.1 | 17 - 17 | 4816 | 131 | |
GenBank mRNA | AB037771.1 | - | 11 - 11 | 3376 | 130 |
AK022799.1 | - | 2 - 2 | 1594 | 130 | |
AK025233.1 | - | 1 - 1 | 1370 | 130 | |
BC017382.1 | - | 1 - 1 | 1257 | 130 | |
BX648339.1 | - | 16 - 16 | 4016 | 131 | |
CR749490.1 | - | 19 - 19 | 5043 | 130 |