Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8122198-8122437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES7
Entrez Gene ID
84667
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
SCDO4, bHLHb37
UniGene
Hs.434828
Gene Chromosome Location
Chr.17: 8120590-8126032 on Build GRCh38
Transcripts
NM_001165967.1 NM_032580.3 XM_011524041.2 XM_017025232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750438 c.142C>T p.L48F Chr.17: 8122427-8122427 on Build GRCh38
5784038 c.147G>T p.R49R Chr.17: 8122422-8122422 on Build GRCh38
4130857 c.175T>C p.L59L Chr.17: 8122394-8122394 on Build GRCh38
5591257 c.206G>A p.R69Q Chr.17: 8122363-8122363 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933336393 Chr.17: 8122396-8122396 on Build GRCh38
rs951701315 Chr.17: 8122260-8122260 on Build GRCh38
rs752215652 Chr.17: 8122309-8122309 on Build GRCh38
rs61758318 Chr.17: 8122326-8122326 on Build GRCh38
rs565314962 Chr.17: 8122222-8122222 on Build GRCh38
rs778723999 Chr.17: 8122340-8122340 on Build GRCh38
rs774091828 Chr.17: 8122306-8122309 on Build GRCh38
rs201434695 Chr.17: 8122391-8122391 on Build GRCh38
rs766087638 Chr.17: 8122303-8122303 on Build GRCh38
rs962844509 Chr.17: 8122224-8122224 on Build GRCh38
rs749037352 Chr.17: 8122322-8122322 on Build GRCh38
rs1025863986 Chr.17: 8122250-8122250 on Build GRCh38
rs756013359 Chr.17: 8122312-8122312 on Build GRCh38
rs387906979 Chr.17: 8122397-8122397 on Build GRCh38
rs751223874 Chr.17: 8122306-8122306 on Build GRCh38
rs773886666 Chr.17: 8122401-8122401 on Build GRCh38
rs117117111 Chr.17: 8122239-8122239 on Build GRCh38
rs370594135 Chr.17: 8122307-8122307 on Build GRCh38
rs569563394 Chr.17: 8122283-8122283 on Build GRCh38
rs994230334 Chr.17: 8122288-8122288 on Build GRCh38
rs1348325 Chr.17: 8122394-8122394 on Build GRCh38
rs747215611 Chr.17: 8122344-8122344 on Build GRCh38
rs768825167 Chr.17: 8122345-8122345 on Build GRCh38
rs866724154 Chr.17: 8122262-8122262 on Build GRCh38
rs547873270 Chr.17: 8122281-8122281 on Build GRCh38
rs914671014 Chr.17: 8122289-8122289 on Build GRCh38
rs781262011 Chr.17: 8122379-8122379 on Build GRCh38
rs1018056328 Chr.17: 8122219-8122219 on Build GRCh38
rs780283648 Chr.17: 8122350-8122350 on Build GRCh38
rs946021143 Chr.17: 8122383-8122383 on Build GRCh38
rs368366222 Chr.17: 8122385-8122385 on Build GRCh38
rs866165247 Chr.17: 8122395-8122395 on Build GRCh38
rs989947309 Chr.17: 8122358-8122358 on Build GRCh38
rs777589200 Chr.17: 8122317-8122317 on Build GRCh38
rs981841965 Chr.17: 8122269-8122269 on Build GRCh38
rs928681827 Chr.17: 8122278-8122278 on Build GRCh38
rs958500760 Chr.17: 8122336-8122336 on Build GRCh38
rs763593387 Chr.17: 8122405-8122405 on Build GRCh38

View Full Product Details