The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025070.1 | NP_001020241.1 | 3 - 3 | 494 | 100 |
NM_001025071.1 | NP_001020242.1 | 3 - 3 | 500 | 100 | |
NM_005617.3 | NP_005608.1 | 3 - 3 | 283 | 100 | |
XM_017009684.1 | XP_016865173.1 | 3 - 3 | 337 | 100 | |
GenBank mRNA | AA467888.1 | - | 1 - 1 | 50 | 100 |
AK312179.1 | - | 3 - 3 | 262 | 100 | |
BC001126.1 | - | 1 - 1 | 246 | 100 | |
BC003401.1 | - | 2 - 2 | 240 | 100 | |
BC006784.1 | - | 1 - 1 | 251 | 100 | |
BC020515.1 | - | 3 - 3 | 433 | 100 | |
BC091474.1 | - | 3 - 3 | 264 | 100 | |
BM454896.1 | - | 3 - 3 | 463 | 100 | |
BM557014.1 | - | 3 - 3 | 435 | 100 |