The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281971.1 | NP_001268900.1 | 4 - 4 | 538 | 150 |
NM_001281972.1 | NP_001268901.1 | 4 - 4 | 538 | 150 | |
GenBank mRNA | AJ417554.1 | - | 3 - 3 | 429 | 150 |
AJ620615.2 | - | 3 - 3 | 422 | 150 | |
AJ620616.2 | - | 3 - 3 | 422 | 150 | |
AY102623.1 | - | 4 - 4 | 449 | 150 | |
AY102624.1 | - | 2 - 2 | 161 | 150 | |
AY366246.1 | - | 4 - 4 | 456 | 150 | |
AY789064.2 | - | 4 - 4 | 433 | 150 |