Product Details

Assay Reference Genome
Location

Chr.8:1783064 on build GRCh38
Cytoband
8p23.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2055
Gene Symbol
CLN8
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
Location
Chr.8:1755778-1786570 on build GRCh38
Assay Gene Location
Within Exon 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CLN8 NM_018941.3 3 3663 NP_061764.2
AF123761.1 3 3592 AAF13119.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1024325 Chr8:651043 - 3203837 on Build GRCh38 Gain LOC101927752 DLGAP2-AS1 MIR596 LOC401442 LOC286083 MIR3674 ERICH1 ARHGEF10 LOC105377792 LOC100131395 LOC105377781 MIR7160 LOC101928058 DLGAP2 LOC105377777 LOC105377776 CSMD1 CLN8 ERICH1-AS1 LOC105377774 LOC101927815 KBTBD11-OT1 KBTBD11 MYOM2
esv3891267 Chr8:1755403 - 2343278 on Build GRCh38 Gain LOC101927752 MIR596 CLN8 MIR3674 ARHGEF10 LOC100131395 LOC105377781 MIR7160 LOC101928058 KBTBD11-OT1 KBTBD11 MYOM2
esv3615841 Chr8:1705527 - 1822229 on Build GRCh38 Loss LOC101927752 MIR596 CLN8 MIR3674 DLGAP2
dgv11822n54 Chr8:1748920 - 1829405 on Build GRCh38 Loss LOC101927752 MIR596 CLN8 MIR3674 ARHGEF10
nsv609527 Chr8:1733567 - 1805331 on Build GRCh38 Loss LOC101927752 CLN8 MIR3674
esv3576504 Chr8:951237 - 1906751 on Build GRCh38 Gain LOC101927752 DLGAP2-AS1 MIR596 LOC286083 CLN8 MIR3674 ERICH1-AS1 ARHGEF10 DLGAP2 LOC105377776
nsv609531 Chr8:1768870 - 2267169 on Build GRCh38 Gain MIR596 CLN8 MIR3674 ARHGEF10 LOC100131395 LOC105377781 MIR7160 LOC101928058 KBTBD11-OT1 KBTBD11 MYOM2
nsv509244 Chr8:1756513 - 1850704 on Build GRCh38 Insertion LOC101927752 MIR596 CLN8 MIR3674 ARHGEF10
nsv1025591 Chr8:1755786 - 2335806 on Build GRCh38 Gain LOC101927752 MIR596 CLN8 MIR3674 ARHGEF10 LOC100131395 LOC105377781 MIR7160 LOC101928058 KBTBD11-OT1 KBTBD11 MYOM2
esv3615846 Chr8:1735209 - 1816717 on Build GRCh38 Loss LOC101927752 CLN8 MIR3674

View Full Product Details