The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039802.1 | NP_001034891.1 | 7 - 7 | 2039 | 102 |
NM_001791.3 | NP_001782.1 | 6 - 6 | 1913 | 102 | |
GenBank mRNA | BC003682.1 | - | 5 - 5 | 1803 | 102 |
BC018266.1 | - | 7 - 7 | 2003 | 102 |