The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136265.1 | NP_001129737.1 | 7 - 8 | 1313 | 75 |
XM_011540630.2 | XP_011538932.1 | 6 - 7 | 1512 | 75 | |
GenBank mRNA | AK024480.1 | - | 3 - 4 | 335 | 75 |