Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFNG
Entrez Gene ID
5986
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
Chromosome Location
Chr.17: 82047902 - 82051774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002917.1 NP_002908.1 7 - 8 924 71
XM_011523587.2 XP_011521889.1 6 - 7 618 71
GenBank mRNA AF038196.1 - 4 - 5 498 71
AK094016.1 - 4 - 5 1139 71
AK293568.1 - 8 - 9 870 71
AK302068.1 - 7 - 8 733 71
BC014495.1 - 3 - 4 372 71
BC069034.1 - 3 - 4 851 71
U94353.1 - 4 - 5 496 71

View Full Product Details