The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020440.3 | NP_065173.2 | 2 - 3 | 701 | 124 |
XM_017001874.1 | XP_016857363.1 | 2 - 3 | 472 | 124 | |
GenBank mRNA | AB037857.1 | - | 2 - 3 | 557 | 124 |
AK074637.1 | - | 2 - 3 | 567 | 124 | |
BC098142.1 | - | 2 - 3 | 147 | 124 | |
BC152454.1 | - | 2 - 3 | 557 | 124 |