The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318046.1 | NP_001304975.1 | 5 - 6 | 489 | 77 |
NM_001318047.1 | NP_001304976.1 | 5 - 6 | 492 | 77 | |
NM_001318048.1 | NP_001304977.1 | 5 - 6 | 489 | 77 | |
NM_007356.2 | NP_031382.2 | 5 - 6 | 492 | 77 | |
XM_011515975.1 | XP_011514277.1 | 5 - 6 | 492 | 77 | |
XM_011515978.1 | XP_011514280.1 | 5 - 6 | 492 | 77 | |
XM_011515979.2 | XP_011514281.1 | 5 - 6 | 492 | 77 | |
XM_011515980.2 | XP_011514282.1 | 5 - 6 | 492 | 77 | |
XM_017011879.1 | XP_016867368.1 | 5 - 6 | 492 | 77 | |
XM_017011880.1 | XP_016867369.1 | 5 - 6 | 492 | 77 | |
GenBank mRNA | AF028816.1 | - | 5 - 6 | 488 | 77 |
AK300385.1 | - | 5 - 6 | 493 | 77 | |
BC140804.1 | - | 5 - 6 | 479 | 77 | |
BC142619.1 | - | 5 - 6 | 474 | 77 | |
BC144591.1 | - | 5 - 6 | 479 | 77 | |
DA859196.1 | - | 5 - 6 | 493 | 77 |