Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMB4
Entrez Gene ID
22798
Gene Name
laminin subunit beta 4
Gene Aliases
-
Chromosome Location
Chr.7: 108011662 - 108130356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC140804.1 - 11 - 12 1432 94
BC142619.1 - 11 - 12 1427 94

View Full Product Details