Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTC28-AS1
- Entrez Gene ID
-
284900
- Gene Name
- TTC28 antisense RNA 1
- Gene Aliases
-
TTC28-AS, TTC28AS, dJ353E16.2
- Chromosome Location
-
Chr.22: 27919376 - 28002679 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026963.1 |
|
3 - 4 |
434 |
105 |
GenBank mRNA |
AB051435.1 |
- |
2 - 3 |
1251 |
105 |
AL832597.1 |
- |
3 - 4 |
389 |
106 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA172828.1 |
- |
2 - 3 |
339 |
|
Chr. 22 - 27919392 - 27997631 [+] on Build GRCh38 |
105 |
GenBankmRNA |
BM921836.1 |
- |
3 - 4 |
685 |
|
Chr. 22 - 27919457 - 27997616 [+] on Build GRCh38 |
105 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 3199-1 |
MIR3199-1 |
mir-3199-1 |
-73.9kb
|
Anti-sense |
Chr. - 27920525 - 27920612 on Build GRCh38 |
Downstream |
microRNA 5739 |
MIR5739 |
hsa-mir-5739 |
462.1kb
|
Sense |
Chr. - 28459869 - 28459948 on Build GRCh38 |
View Full Product Details