Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC153684
- Entrez Gene ID
-
153684
- Gene Name
- uncharacterized LOC153684
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 43042134 - 43045268 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM555934.1 |
- |
2 - 3 |
168 |
|
Chr. 5 - 43041807 - 43045281 [+] on Build GRCh38 |
98 |
GenBankmRNA |
BU942263.1 |
- |
1 - 2 |
244 |
|
Chr. 5 - 43043330 - 43044783 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC648987 |
LOC648987 |
|
|
Anti-sense |
Chr. - 43014729 - 43067439 on Build GRCh38 |
Downstream |
uncharacterized LOC100132356 |
LOC100132356 |
|
19.9kb
|
Anti-sense |
Chr. - 43065187 - 43066971 on Build GRCh38 |
View Full Product Details