Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr.21: 31671000 - 31732118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145444.1 NP_001138916.1 1 - 2 459 76
NM_001145445.1 NP_001138917.1 1 - 2 459 76
NM_020706.2 NP_065757.1 1 - 2 459 76
XM_005261017.2 XP_005261074.1 1 - 2 440 76
XM_006724035.2 XP_006724098.1 1 - 2 440 76
XM_006724036.2 XP_006724099.1 1 - 2 440 76
XM_017028415.1 XP_016883904.1 1 - 2 420 76
XM_017028416.1 XP_016883905.1 1 - 2 305 76
XM_017028417.1 XP_016883906.1 1 - 2 440 76
GenBank mRNA AK057840.1 - 1 - 2 302 76
AK308406.1 - 1 - 2 309 76
AL834304.1 - 1 - 2 239 76
BC014921.1 - 1 - 2 436 76
BC043353.1 - 1 - 2 416 76
BC052286.1 - 1 - 2 290 76
BC064990.1 - 1 - 2 290 76

View Full Product Details