The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145444.1 | NP_001138916.1 | 8 - 9 | 1446 | 71 |
NM_001145445.1 | NP_001138917.1 | 9 - 10 | 1491 | 71 | |
NM_020706.2 | NP_065757.1 | 9 - 10 | 1491 | 71 | |
XM_005261017.2 | XP_005261074.1 | 8 - 9 | 1295 | 71 | |
XM_006724035.2 | XP_006724098.1 | 9 - 10 | 1472 | 71 | |
XM_006724036.2 | XP_006724099.1 | 9 - 10 | 1472 | 71 | |
XM_017028415.1 | XP_016883904.1 | 9 - 10 | 1449 | 71 | |
XM_017028416.1 | XP_016883905.1 | 9 - 10 | 1334 | 71 | |
XM_017028417.1 | XP_016883906.1 | 8 - 9 | 1295 | 71 | |
GenBank mRNA | AB032998.1 | - | 3 - 4 | 479 | 71 |
AF023142.1 | - | 9 - 10 | 1095 | 71 | |
AK025748.1 | - | 4 - 5 | 510 | 71 | |
AK308406.1 | - | 8 - 9 | 1297 | 71 | |
AL117417.1 | - | 4 - 5 | 499 | 71 | |
AL834304.1 | - | 9 - 10 | 1271 | 71 | |
BC014921.1 | - | 9 - 10 | 1468 | 71 | |
BC043353.1 | - | 9 - 10 | 1448 | 71 | |
BC052286.1 | - | 9 - 10 | 1322 | 71 | |
BC064990.1 | - | 9 - 10 | 1322 | 71 |