Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
Chromosome Location
Chr.Y: 6865918 - 6874056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143.1 NP_001134.1 1 - 2 58 91
XM_017030042.1 XP_016885531.1 1 - 2 87 91
GenBank mRNA BC069138.1 - 1 - 2 58 91
M86933.1 - 1 - 2 58 91

View Full Product Details