Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
-
Chromosome Location
Chr.9: 129612225 - 129622275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199350.3 NP_955382.3 4 - 5 1079 69
XM_011518655.1 XP_011516957.1 6 - 7 1634 69
XM_011518656.1 XP_011516958.1 5 - 6 1144 69
XM_011518657.2 XP_011516959.1 5 - 6 875 69
XM_011518658.2 XP_011516960.1 5 - 6 872 69
XM_011518661.2 XP_011516963.1 2 - 3 608 69
XM_011518662.2 XP_011516964.1 2 - 3 605 69
GenBank mRNA AK093122.1 - 4 - 5 1079 69
AW958564.1 - 1 - 2 52 69
AW958565.1 - 1 - 2 52 69
BC112353.1 - 2 - 3 299 69
JF442034.1 - 4 - 5 69

View Full Product Details