The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013841.1 | NP_001013863.1 | 10 - 11 | 1080 | 62 |
NM_017720.2 | NP_060190.2 | 10 - 11 | 1080 | 62 | |
XM_011528123.1 | XP_011526425.1 | 10 - 11 | 1107 | 62 | |
GenBank mRNA | AJ245719.1 | - | 10 - 11 | 1041 | 62 |
AK000241.1 | - | 10 - 11 | 1051 | 62 | |
AK125774.1 | - | 12 - 13 | 1590 | 62 | |
BC000795.1 | - | 10 - 11 | 1031 | 62 |