Product Details

Assay Reference Genome
Location

Chr.5:78002925 on build GRCh38
Cytoband
5q14.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8546
Gene Symbol
AP3B1
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
Location
Chr.5:78002326-78294755 on build GRCh38
Assay Gene Location
Within Exon 29
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
AP3B1 NM_001271769.1 27 3450 NP_001258698.1
NM_003664.4 27 3438 NP_003655.3
XM_005248618.3 28 3446 XP_005248675.1
XM_017010001.1 28 3226 XP_016865490.1
AK225908.1 27 3314
AK291201.1 27 3379
BC038444.1 27 3399 AAH38444.1
BX538041.1 27 3387 CAD97982.1
KU178426.1
KU178427.1
KU178428.1
U81504.1 27 3354 AAB61638.1
U91931.1 27 3312 AAD03778.1

View Full Product Details