The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139442.1 | NP_001132914.1 | 1 - 2 | 921 | 71 |
NM_194252.2 | NP_919228.2 | 1 - 2 | 921 | 71 | |
XM_005251728.2 | XP_005251785.1 | 1 - 2 | 356 | 71 | |
XM_006716966.3 | XP_006717029.1 | 1 - 2 | 598 | 71 | |
XM_017014309.1 | XP_016869798.1 | 1 - 2 | 598 | 71 | |
GenBank mRNA | AF521886.1 | - | 1 - 2 | 921 | 71 |
AF521887.1 | - | 1 - 2 | 81 | 71 | |
AK125178.1 | - | 1 - 2 | 599 | 71 |