Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LONRF2
Entrez Gene ID
164832
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
RNF192
Chromosome Location
Chr.2: 100273291 - 100322733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198461.3 NP_940863.3 10 - 11 2561 69
GenBank mRNA AK127206.1 - 10 - 11 1263 69
BC101662.1 - 9 - 10 1220 69
BC101664.1 - 9 - 10 1220 69
BC143468.1 - 9 - 10 1220 69

View Full Product Details