Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFYVE19
Entrez Gene ID
84936
Gene Name
zinc finger FYVE-type containing 19
Gene Aliases
ANCHR, MPFYVE
Chromosome Location
Chr.15: 40807076 - 40814569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017022683.1 XP_016878172.1 1 - 1 637 147
GenBank mRNA AB209224.1 - - 1227 147

View Full Product Details