The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276464.1 | NP_001263393.1 | 2 - 3 | 259 | 100 |
NM_003822.4 | NP_003813.1 | 2 - 3 | 428 | 100 | |
NM_205860.2 | NP_995582.1 | 3 - 4 | 566 | 100 | |
XM_005245062.3 | XP_005245119.1 | 2 - 3 | 693 | 100 | |
XM_011509380.1 | XP_011507682.1 | 4 - 5 | 1154 | 100 | |
XM_011509381.2 | XP_011507683.1 | 3 - 4 | 2018 | 100 | |
XM_011509382.1 | XP_011507684.1 | 3 - 4 | 1016 | 100 | |
XM_011509384.2 | XP_011507686.1 | 3 - 4 | 399 | 100 | |
XM_017000904.1 | XP_016856393.1 | 2 - 3 | 271 | 100 | |
GenBank mRNA | AB019246.1 | - | 3 - 4 | 446 | 100 |
AB307719.1 | - | 2 - 3 | 200 | 100 | |
AF049102.1 | - | 1 - 2 | 82 | 100 | |
AF124247.1 | - | 3 - 4 | 320 | 100 | |
AF146343.1 | - | 2 - 3 | 389 | 100 | |
AK304365.1 | - | 2 - 3 | 259 | 100 | |
AK308446.1 | - | 2 - 3 | 382 | 100 | |
AK316513.1 | - | 2 - 3 | 495 | 100 | |
BC118571.1 | - | 2 - 3 | 275 | 100 | |
BC118652.1 | - | 2 - 3 | 275 | 100 | |
HQ692857.1 | - | 2 - 3 | 182 | 100 | |
U80251.1 | - | 2 - 3 | 237 | 100 | |
U93553.1 | - | 3 - 4 | 562 | 100 |